missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Guanidine Hydrochloride, ≥99%, ULTROL™ Grade, Calbiochem™,
Supplier: MilliporeSigma™ 3690751KG
| Quantity | 1 kg |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| Guanidinium Chloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
Specifications
| White | |
| 1 kg | |
| CH6ClN3 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| ≥99% (Titration) |
| 50-01-1 | |
| Glass Bottle | |
| Guanidinium Chloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 | |
| ULTROL |