missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Guanidine Hydrochloride, ≥99%, ULTROL™ Grade, Calbiochem™,

Supplier:  MilliporeSigma™ 369075100GM

 View more versions of this product

Catalog No. 369075100GM


Only null left

Description

Description

Specifications

Specifications

White
100 g
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
≥99% (Titration)
50-01-1
Glass Bottle
Guanidinium Chloride
C(=N)(N)N.Cl
95.53
CHEBI:32735
ULTROL

Chemical Identifiers

50-01-1
95.53
Guanidinium Chloride
CHEBI:32735
C(=N)(N)N.Cl
CH6ClN3
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions