Learn More
Guanidine Hydrochloride, >99.5%, Molecular Biology Grade
CAS: 50-01-1 | CH6ClN3 | 95.53 g/mol
Supplier: Thermo Fisher Scientific J75823A1
Description
- Used for extraction of RNA.
- Preparation of high molecular weight genomic DNA.
- Solubilizing inclusion bodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Used for extraction of RNA
- Preparation of high molecular weight genomic DNA
- Solubilizing inclusion bodies
Specifications
| Guanidine hydrochloride | |
| 185°C to 188°C | |
| Plastic bottle | |
| (NH2)2C=NH.HCl | |
| 1 kg | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 | |
| >99.5% | |
| Crystals |
| 50-01-1 | |
| 1.344 | |
| CH6ClN3 | |
| MFCD00013026 | |
| 3591990 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| 95.53 | |
| Molecular Biology |
Chemical Identifiers
| 50-01-1 | |
| 95.53 | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| 5742 | |
| guanidine;hydrochloride |
| CH6ClN3 | |
| MFCD00013026 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| CHEBI:32735 | |
| C(=N)(N)N.Cl |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H500
EINECSNumber : 200-002-3
RTECSNumber : MF4300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only