missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanidine Hydrochloride, ≥99.5%, (Crystalline/Ultra Pure), MP Biomedicals
Specifications
Specifications
| Assay Percent Range | ≥99.5% |
| Quantity | 25 g |
| Formula Weight | 95.5 |
| Grade | Ultrapure |
| Physical Form | Crystalline |
| Chemical Name or Material | Guanidine Hydrochloride |
Safety and Handling
| Recommended Storage | Room Temperature |