missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrochloride, ≥99.5%, (Crystalline/Ultra Pure), MP Biomedicals

Catalog No. ICN10569625
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10569625 25 g
ICN10569680 100 g
ICN10569690 500 g
ICN10569691 1 kg
ICN10569694 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10569625 Supplier MP Biomedicals Inc Supplier No. 0210569625

Forms clear, colorless solutions at 6.0 M concentration. O.D. 260 nm of a 6.0 M solution: <0.03

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH6ClN3
Molecular Weight (g/mol) 95.53
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl

Specifications

Assay Percent Range ≥99.5%
Quantity 25 g
Formula Weight 95.5
Grade Ultrapure
Physical Form Crystalline
Chemical Name or Material Guanidine Hydrochloride
Recommended Storage Room Temperature