Learn More
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Thermo Scientific™
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), ACROS Organics, Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry)
Supplier: Thermo Scientific Chemicals 120230100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Guanidine hydrochloride | |
| 1.3400g/mL | |
| 0.5% max. Ammonium chloride (Argentometry) | |
| CH5N3·HCl | |
| 10kg | |
| 1.34 | |
| guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride | |
| PJJJBBJSCAKJQF-UHFFFAOYSA-N | |
| guanidine;hydrochloride | |
| 5742 | |
| 95.53 |
| 50-01-1 | |
| Authentic | |
| Plastic drum | |
| H2NC(=NH)NH2·HCl | |
| 03,82 | |
| 15,4597 | |
| Solubility in water: 2280g/L (20°C). Other solubilities: 300g/L ethanol (30°C), 19.5g/100g dimethylformamide (30°C), <0.05g/100g acetone -<0.05g/100g benzene (30°C), insoluble in trichloroethylene | |
| C(=N)(N)N.Cl | |
| 95.53 | |
| CHEBI:32735 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Warning
EINECSNumber : 200-002-3