missing translation for 'onlineSavingsMsg'
Learn More

Guanidine Hydrobromide 98.0+%, TCI America™

Catalog No. G04495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04495G 5 g
G04491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04495G Supplier TCI America Supplier No. G04495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19244-98-5
Molecular Formula CH6BrN3
Molecular Weight (g/mol) 139.984
MDL Number MFCD03094062
InChI Key VQNVZLDDLJBKNS-UHFFFAOYSA-N
Synonym GABr, Guanidinium Bromide
PubChem CID 71282
IUPAC Name guanidine;hydrobromide
SMILES C(=N)(N)N.Br

Specifications

Melting Point 191°C
Color White
Quantity 5 g
Formula Weight 139.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Guanidine Hydrobromide