accessibility menu, dialog, popup

UserName

CAS RN 19244-98-5

IUPAC Name: hydrogen guanidine bromide
Synonyms: hydrogen guanidine bromide
Molecular Weight (g/mol): 139.98
Molecular Formula: CH6BrN3
InChi Key: VQNVZLDDLJBKNS-UHFFFAOYSA-N
SMILES: [H+].[Br-].NC(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)