missing translation for 'onlineSavingsMsg'
Learn More

Guanfacine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013834
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013834 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013834 Supplier Merck Emd Millipore Supplier No. PHR1751500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Guanfacine hydrochloride, is a selective α-adrenergic receptor agonist, that is used in the treatment of attention deficit hyperactivity disorder (ADHD) and hypertension.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 29110-48-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H9 Cl2N3 O · HCl
MDL Number MFCD00798230
Synonym Guanfacine hydrochloride; N (Aminoiminomethyl)-2,6-dichlorobenzeneacetamide
Type Certified Reference Material
Quantity 500 mg
Formula Weight 282.55
Grade Certified Reference Material