missing translation for 'onlineSavingsMsg'
Learn More

Guanethidine sulfate, Thermo Scientific™

Catalog No. AC468622500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468622500 250 mg
AC468620010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468622500 Supplier Thermo Scientific Chemicals Supplier No. 468622500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 645-43-2
Molecular Formula C10H24N4O4S
Molecular Weight (g/mol) 296.39
MDL Number MFCD00035403
InChI Key YUFWAVFNITUSHI-UHFFFAOYSA-N
IUPAC Name N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid
SMILES OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Concentration or Composition Notes Titration with HClO4
Physical Form Crystalline Powder/Crystals
Concentration 97.5% to 102.5%
Color White to Light Beige
Quantity 250 mg
Formula Weight 296.39
Chemical Name or Material Guanethidine sulfate
Hazard Category GHS07: Exclamation mark
ShelfLife 2 years
EINECSNumber 211-442-0
RTECSNumber MF3150000
Recommended Storage Normal conditions