missing translation for 'onlineSavingsMsg'
Learn More

Guanabenz Acetate 98.0+%, TCI America™

Catalog No. G0456200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0456200MG 200 mg
G04561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0456200MG Supplier TCI America Supplier No. G0456200MG
Only null left

Chemical Identifiers

CAS 23256-50-0
Molecular Formula C10H12Cl2N4O2
Molecular Weight (g/mol) 291.132
MDL Number MFCD00153801
InChI Key MCSPBPXATWBACD-UHFFFAOYSA-N
Synonym guanabenz wy-8678 acetate, 2,6-dichlorobenzylidene-amino guanidine, acetic acid; 2-2,6-dichlorophenyl methylideneamino guanidine, n-2,6-dichlorophenyl methylidene amino guanidine; acetic acid
PubChem CID 6852405
IUPAC Name acetic acid;2-[(2,6-dichlorophenyl)methylideneamino]guanidine
SMILES CC(=O)O.C1=CC(=C(C(=C1)Cl)C=NN=C(N)N)Cl

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 291.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Guanabenz Acetate
RTECSNumber MF0382000
TSCA No