accessibility menu, dialog, popup

UserName

CH3OHONH2H2NNNClCl(E)

CAS RN 23256-50-0

IUPAC Name: N''-[(E)-[(2,6-dichlorophenyl)methylidene]amino]guanidine; acetic acid
Synonyms: acetic acid; guanabenz
Molecular Weight (g/mol): 291.13
Molecular Formula: C10H12Cl2N4O2
InChi Key: MCSPBPXATWBACD-GAYQJXMFSA-N
SMILES: CC(O)=O.NC(N)=N\N=C\C1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results