missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Guaifenesin, USP
Description
Guaifenesin USP is a white to slightly gray, crystalline powder. May have a slight characteristic odor.
Alternative name(s): GLycerol guaiacolate, guaiphenesin
Specifications
Specifications
| Chemical Name or Material | Guaifenesin, USP |
| Packaging | Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT |
| CAS | 93-14-1 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 198.2166 |
| Quantity | 500 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |