missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol 98.0+%, TCI America™

Catalog No. p-7132003
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012125G 25 g
M0121500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M012125G Supplier TCI America Supplier No. M012125G

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002185
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Color White
Boiling Point 205°C
Quantity 25 g
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Guaiacol
EINECSNumber (3)-0567
RTECSNumber SL7525000
TSCA Yes