missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol, Purified, 99%, Spectrum™ Chemical

Catalog No. 18603066
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603066 100 mL Amber Glass Bottle
18603067 500 mL Amber Glass Bottle
18603068 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603066 Supplier Spectrum Chemical Mfg Cor Supplier No. GU101100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

GU101, 90-05-1, C7H8O2

Spectrum™ Chemical Guaiacol, Purified is a natural organic compound that is biosynthesized by several different organisms. It is a yellow pleasant smelling oil derived most commonly from wood creosote. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
Percent Purity 99%
Grade Purified
Physical Form Liquid