missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015826
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015826 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015826 Supplier Merck Emd Millipore Supplier No. PHR11361.5G

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 90-05-1
Vapor Pressure 0.11 mmHg (25°C)
Vapor Density 4.27 (vs. air)
Density 1.129 g/mL (at 25°C)
Flash Point 82°C - Closed Cup
Linear Formula (CH3 O)C6H4 OH
MDL Number MFCD00002185
Melting Point 26°C to 29°C
Boiling Point 205°C
Synonym 2-Methoxyphenol; Catechol monomethyl ether; Pyrocatechol monomethyl ether
Type Certified Reference Material
Quantity 5 g
Formula Weight 124.14
Grade Certified Reference Material
Show More Show Less