missing translation for 'onlineSavingsMsg'
Learn More

Guaiacol Glycerol Ether 98.0+%, TCI America™

Catalog No. p-7130168
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
G015925G 25 g
G0159500G 500 g
2 options

Catalog No. G015925G

Supplier: TCI America G015925G

Only null left

Chemical Identifiers

CAS 93-14-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00016873
InChI Key HSRJKNPTNIJEKV-UHFFFAOYNA-N
Synonym guaifenesin, guaiphenesin, glyceryl guaiacolate, glycerol guaiacolate, guaiphenesine, guaiacol glyceryl ether, methphenoxydiol, aeronesin, breonesin, bronchol
PubChem CID 3516
IUPAC Name 3-(2-methoxyphenoxy)propane-1,2-diol
SMILES COC1=CC=CC=C1OCC(O)CO

Specifications

Melting Point 83°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 198.22
Percent Purity ≥98.0% (HPLC,E)
Physical Form Crystalline Powder
Chemical Name or Material Guaiacol Glycerol Ether
EINECSNumber (3)-0576&(3)-0588
RTECSNumber TY8400000
TSCA Yes