missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GTSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558575 Supplier Invitrogen™ Supplier No. PA558575
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: QTRSLRQETL AESTWQCPPC DEDWDKDLWE QTSTPFVWGT THYSDNNSPA SNIVTEHKNN LASGMRVPKS LPYV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

Specifications

Antigen GTSF1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTSF1
Gene Accession No. Q8WW33
Gene Alias 1700006H03Rik; Cue110; Fam112b; family with sequence similarity 112, member B; gametocyte specific factor 1; gametocyte-specific factor 1; Gtsf1; Protein FAM112B; RGD1310791
Gene Symbols GTSF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTSF1. Recombinant protein control fragment (Product #RP-97832).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121355
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less