missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF3C3/TFIIIC102 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
RNA polymerase (pol) III synthesizes tRNA, 5s rRNA, 7SL RNA and U6 snRNA and is overexpressed in many transformed cell lines and tumors in vivo, since cells must duplicate its protein components before division. Therefore, in order to maintain rapid growth, cells must produce a high level of Pol III transcribed RNA, which requires the presence of the TFIIIB and TFIIIC2 transcription factor complexes. The TFIIIC2 complex is composed of five subunits, TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, that are overexpressed in adenovirus transformed cells as well as in maligt cells in vivo, such as ovarian carcinomas. TFIIIC2 recruits RNA pol III and TFIIIB to promoter elements and may be a key component in the deregulation of maligt cells. The TFIIIB complex includes the TATA-binding protein (TBP), TFIIB-related factor 1 (BRF1) and TFIIIB, the expression of which are also upregulated in transformed cells. In many carcinomas, the tumor suppressors retinoblastoma (RB) and p53 are inactivated, which affects their ability to bind and inactivate the function of TFIIIB.
Specifications
Specifications
| Antigen | GTF3C3/TFIIIC102 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | GTF3C3 |
| Gene Accession No. | Q9Y5Q9 |
| Gene Alias | General transcription factor 3C polypeptide 3, Transcription factor IIIC 102 kDa subunit, TFIIIC 102 kDa subunit, TFIIIC102, Transcription factor IIIC subunit gamma, TF3C-gamma, GTF3C3 |
| Gene Symbols | Gtf3c3 |
| Host Species | Rabbit |
| Show More |