missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT2B Monoclonal Antibody (K24025_5C8), Invitrogen™
Mouse Monoclonal Antibody
$1421.98 - $2134.69
Specifications
| Antigen | GSTT2B |
|---|---|
| Clone | K24025_5C8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: SQPSRAVYIF AKKNGIPLEL RTVDLVKGQH KSKEFLQINS LGKLPTLKDG DFILTESSAI LIYLSCKYQT PDHWYPSDLQ ARARVHEYLG WHADCIRGTF GIPLWVQVLG PLIGVQVPEE KVERNRTAMD QALQWLEDKF LGDRPFLAGQ QVTLADLMAL EELMQPVALG YELFEGRPRL AAWRGRVEAF LGAELCQEAH SIILSILEQA AKKTLPTPSP
GSTT2B Monoclonal antibody specifically detects GSTT2B in Human, Rat samples. It is validated for Western BlotSpecifications
| GSTT2B | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| glutathione S-transferase theta 2; glutathione S-transferase theta 2B (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase theta-2B; GST class-theta-2; GSTT2; GSTT2B; GSTT2P | |
| Recombinant human GSTT2B (immunogen range is 11-230). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GSTT2B |
| K24025_5C8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P0CG30 | |
| 653689 | |
| IgG2a | |
| Protein A | |
| Antibody |