missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSTP1 Monoclonal Antibody (K52024_5E8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555980
Description
Sequence of this protein is as follows: MPPYTVVYFP VRGRCAALRM LLADQGQSWK EEVVTVETWQ EGSLKASCLY GQLPKFQDGD LTLYQSNTIL RHLGRTLGLY GKDQQEAALV DMVNDGVEDL RCKYISLIYT NYEAGKDDYV KALPGQLKPF ETLLSQNQGG KTFIVGDQIS FADYNLLDLL LIHEVLAPGC LDAFPLLSAY VGRLSARPKL KAFLASPEYV NLPINGNGKQ
Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.Specifications
| GSTP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P09211 | |
| Gstp1 | |
| Recombinant human GSTP1 (immunogen range is 1-210). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K52024_5E8 | |
| Unconjugated | |
| Gstp1 | |
| Chain 7; deafness, X-linked 7; DFN7; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-transferase P; Glutathione S-transferase P 1; glutathione S-transferase pi 1; glutathione S-transferase pi 2; glutathione S-transferase, pi 1; glutathione S-transferase, pi 2; Glutathione-S-transferase placental enzyme pi type; glutathione-S-transferase, pi 1; Glutathione-S-transferase, placental enzyme pi type; GST 7-7; GST class-pi; gst P1; Gst p-1; GST YF-YF; Gst3; Gstp; GST-P; Gstp1; GSTP1-1; Gstp2; GstpiB; GST-piB; HEL-S-22; HGNC:4638; PI; Preadipocyte growth factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 2950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |