missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GSTCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561167 Supplier Invitrogen™ Supplier No. PA561167
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: TRASFVTCPC CYGFIQNTSK FNFPKSEQFK KTLSYKEHMI LCRFADQTAV QLPPQRRLIG KQCMCLVDLD RARAAEECGY SVQVI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

GSTCD (glutathione S-transferase, C-terminal domain containing) is a 633 amino acid protein belonging to the GSTCD family and contains one GST C-terminal domain. The gene encoding GSTCD maps to human chromosome 4, which represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer.

Specifications

Antigen GSTCD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gstcd
Gene Accession No. Q8NEC7
Gene Alias 4933434L15Rik; glutathione S-transferase C-terminal domain containing; glutathione S-transferase C-terminal domain-containing protein; glutathione S-transferase, C-terminal domain containing; GSTCD; RGD1311822
Gene Symbols Gstcd
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GSTCD. Recombinant protein control fragment (Product #RP-96665).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less