missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSS Recombinant Rabbit Monoclonal Antibody (23GB4385)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA554231
Description
GSS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Glutathione is important for a variety of biological functions, including protection of cells from oxidative damage by free radicals, detoxification of xenobiotics, and membrane transport. The protein encoded by this gene functions as a homodimer to catalyze the second step of glutathione biosynthesis, which is the ATP-dependent conversion of gamma-L-glutamyl-L-cysteine to glutathione. Defects in this gene are a cause of glutathione synthetase deficiency.Specifications
| GSS | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P46413, P48637, P51855 | |
| GSS | |
| A synthesized peptide derived from human Glutathione Synthetase (321-400AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4385 | |
| Unconjugated | |
| GSS | |
| AI314904; Glutathione synthase; glutathione synthetase; GS-A/GS-B; GSH S; GSH synthetase; GSHB; GSHS; GSH-S; GSS; HEL-S-64p; HEL-S-88n; MGC14098; OTTHUMP00000030711 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14854, 25458, 2937 | |
| -20°C | |
| Liquid |