missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSK3 alpha/beta Recombinant Rabbit Monoclonal Antibody (23GB1100)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA554075
Description
GSK3 alpha/beta Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Glycogen synthase kinase-3 (GSK3) is a protein kinase that was originally identified as a regulator of glycogen synthase, a key enzyme in glycogen metabolism. Since then, it has been shown to be involved in the regulation of a diverse array of cellular functions, including protein synthesis, cell proliferation, cell differentiation, microtubule assembly/disassembly, and apoptosis. GSK3's substrate specificity is unique in that phosphorylation of the substrate only occurs if a phosphoserine or phosphotyrosine is present - four residues C-terminal to the site of GSK phosphorylation. There exists two isoforms of GSK3, GSK3 alpha and GSK3 beta, and they are strictly regulated via phosphorylation. Phosphorylation of GSK3 beta on Ser9 (Ser21 in GSK3alpha) by protein kinase B (PKB) causes its inactivation and is the primary mechanism responsible for growth factor inhibition of this kinase. Activation of GSK3 beta is dependent upon the phosphorylation of Tyr216 (Tyr279 in GSK3 alpha). Upon activation, it has been shown to phosphorylate a number of different cellular proteins, including p53, c-Myc, c-Jun, heat shock factor-1 (HSF-1), and cyclin D1. GSK3 beta also has been shown to phosphorylate aberrant sites on the microtubule associated protein tau, which is critical for the progression of Alzheimer's disease.Chemical Identifiers
| GSK3 alpha/beta | |
| 0.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18265, P18266, P49840, P49841, Q2NL51, Q9WV60 | |
| 2931, 2932, 50686, 56637, 606496, 84027 | |
| Recombinant protein of human GSK3 alpha (272-283AA). | |
| Antibody |
| 23GB1100 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GSK3A | |
| 2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B | |
| GSK3A, GSK3B | |
| Primary | |
| Affinity Chromatography |
Specifications
| GSK3 alpha/beta | |
| Recombinant Monoclonal | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18265, P18266, P49840, P49841, Q2NL51, Q9WV60 | |
| GSK3A, GSK3B | |
| Recombinant protein of human GSK3 alpha (272-283AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1100 | |
| Unconjugated | |
| GSK3A | |
| 2700086H06Rik; 7330414F15Rik; 8430431H08Rik; C86142; FA; factor A; glycogen synthase kinase 3 alpha; glycogen synthase kinase 3 beta; glycogen synthase kinase-3 alpha; Glycogen synthase kinase-3 beta; GSK3; GSK-3; GSK-3 alpha; GSK-3 beta; Gsk3a; gsk-3-alpha; Gsk3b; GSK-3beta; GSK3beta isoform; serine/threonine-protein kinase GSK3A; Serine/threonine-protein kinase GSK3B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2931, 2932, 50686, 56637, 606496, 84027 | |
| -20°C | |
| Liquid |