missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals GSK-3beta Inhibitor VIII

Catalog No. aaj64554ma
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64554MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64554MA Supplier Thermo Scientific Chemicals Supplier No. J64554MA

CAS: 487021-52-3 | C12H12N4O4S, C12H12N4O4S | 308.31 g/mol

A cell-permeable, potent, and selective inhibitor of GSK-3beta

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 487021-52-3
Molecular Formula C12H12N4O4S
Molecular Weight (g/mol) 308.31
InChI Key YAEMHJKFIIIULI-UHFFFAOYSA-N
Synonym AR-A014418
IUPAC Name 1-[(4-methoxyphenyl)methyl]-3-(5-nitro-1,3-thiazol-2-yl)urea
SMILES COC1=CC=C(CNC(=O)NC2=NC=C(S2)[N+]([O-])=O)C=C1

Specifications

Quantity 10 mg
Description Yellow
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, potent, and selective inhibitor of GSK-3beta
Formula Weight 308.31g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Solid
Recommended Storage Keep cold

RUO – Research Use Only