Learn More
MilliporeSigma™ Ground Joint Flasks for Vacuum Filtration
Accessories for filter holders for sample preparation.
$107.00 - $3901.41
| Catalog Number | Mfr. No. | Volume (Metric) | Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Type | Price | Quantity | ||||
XX1614705
|
MilliporeSigma™
XX1614705 |
1 L | Conical Bottom Flask |
Each for $2,144.23
|
|
||||
XX1614706
|
MilliporeSigma™
XX1614706 |
5 L | Conical Bottom Flask |
Each for $3,901.41
|
|
||||
|
XX1514705
|
MilliporeSigma™
XX1514705 |
1 L | Standard Bottom Flask |
Pack for $451.00
|
|
||||
|
XX7100004
|
MilliporeSigma™
XX7100004 |
250 mL | Tubing |
Each for $107.00
|
|
||||
|
XX1014745
|
MilliporeSigma™
XX1014745 |
4 L | Standard Bottom Flask |
Each for $912.58
|
|
||||
Description
- These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
- This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.