missing translation for 'onlineSavingsMsg'
Learn More

SPEX™ SamplePrep Grinding Vial Accessory Packages

Catalog No. 6870S
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
6870S Small
6870L Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6870S Supplier SPEX™ SamplePrep Supplier No. 6870S

Contains everything necessary to run Small/Mid-size/Large grinding vials in the LargeFreezer/Mill™

  • Closed grinding vial prevents cross-contamination.
  • Excellent for grinding tissue, seeds, polymers, pharmaceuticals, and textiles.
  • Compatible with grinding vial sets 04576-35 and 61043-07.
  • Compatible with Freezer/Mill™ cryogenic grinders 04500-39 to -42, 04577-77, and 04577-92 to -94.

Specifications

Product Type Grinding Set
For Use With (Equipment) Freezer/Mill Cryogenic Grinders
Includes 1 Multivial Holder (04500-51), 1 Vial Extractor/Opener (04576-31), and 1 Vial Rack (04576-32)
Size Small