missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRHL1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA552293

Catalog No. PIPA552293

Only null left
Explore available promotions

Description

Description

Immunogen sequence: VTVSIATMPT HSIKTETQPH GFAVGIPPAV YHPEPTERVV VFDRNLNTDQ FSSGAQAPNA QRRTPDSTFS ETFKEGVQEV FFPSDLSLRM PGMNSEDYVF DSVSGNNFEY TLEASKSLRQ KPGDSTMTYL NK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

This gene encodes a member of the grainyhead family of transcription factors. The encoded protein can exist as a homodimer or can form heterodimers with sister-of-mammalian grainyhead or brother-of-mammalian grainyhead. This protein functions as a transcription factor during development.
Specifications

Specifications

GRHL1
Polyclonal
Unconjugated
GRHL1
DNA methyltransferase 3A; Dnmt3a; grainyhead like 1; grainyhead like transcription factor 1; grainyhead-like 1; grainyhead-like 1 (Drosophila); grainyhead-like protein 1 homolog; grainyhead-like transcription factor 1; GRHL1; LBP protein 32; LBP32; LBP-32; Mammalian grainyhead; Mgr; NH32; p61 MGR; p70 MGR; Tcfcp2l2; TFCP2L2; transcription factor CP2-like 2; Transcription factor LBP-32
Rabbit
Antigen affinity chromatography
RUO
29841
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NZI5
GRHL1
Recombinant protein corresponding to Human GRHL1. Recombinant protein control fragment (Product #RP-89349).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions