Learn More
Qorpak™ Green Thermoset F217 and PTFE Lined Caps
Offer wide range of chemical compatibility and high temperature tolerance
Supplier:  Qorpak™ CAP00061
 View more versions of this product
Catalog No. 361027770
Only null left
Description
Description
  • Resist attack from chemicals at room temperature
  • Can be used when odor and impact resilient tight seal is required
  • With low moisture transmission rate
  • Available in ribbed and smooth styles
  • Available in bagged or bulk packing
Specifications
Chemical Identifiers
Green
Non-sterile
Reusable
Wide mouth bottle
Not Autoclavable
Thermoset
F217, PTFE
Threaded Screw Cap
Smooth
Specifications
Green
144 Cs.
Bagged
F217, PTFE
Reusable
Wide mouth bottle
Not Autoclavable
Thermoset
12/Pk.
Non-sterile
83-400
Threaded Screw Cap
Smooth
Product Suggestions
Product Suggestions