missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ Green Thermoset F217 and PTFE Lined Caps

Catalog No. p-4603612
Change view
Click to view available options
Neck Finish:
43-400
45-400
48-400
53-400
58-400
63-400
70-400
83-400
89-400
No. per Case:
1400 Cs.
144 Cs.
288 Cs.
340 Cs.
500 Cs.
858 Cs.
Packaging:
Bagged
Bulk
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02993408 45-400 500 Cs. Bagged
361027770 83-400 144 Cs. Bagged
02993420 89-400 144 Cs. Bagged
02991028 58-400 858 Cs. Bulk
02993407 48-400 500 Cs. Bagged
02991012 48-400 1400 Cs. Bulk
361010390 83-400 340 Cs. Bulk
02993409 53-400 500 Cs. Bagged
02993412 43-400 500 Cs. Bagged
02993411 63-400 288 Cs. Bagged
361010373 58-400 500 Cs. Bagged
02993430 70-400 144 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 02993408 Supplier Qorpak™ Supplier No. CAP00068
Only null left

Offer wide range of chemical compatibility and high temperature tolerance

  • Resist attack from chemicals at room temperature
  • Can be used when odor and impact resilient tight seal is required
  • With low moisture transmission rate
  • Available in ribbed and smooth styles
  • Available in bagged or bulk packing

Specifications

Color Green
Material Thermoset
No. per Case 500 Cs.
No. per Pack 100/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material F217, PTFE
Neck Finish 45-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Closure Type Smooth
Autoclavable Not Autoclavable
Show More Show Less