missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Green S, ≥98% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 3087-16-9 |
| Linear Formula | C27H25N2 NaO7S2 |
| MDL Number | MFCD00012117 |
| Synonym | Acid Brilliant Green BS; Acid Green 50; Wool Green S |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 576.62 |
| Percent Purity | ≥98% (HPLC) |
| Grade | Analytical Standard |