missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRB7 Recombinant Rabbit Monoclonal Antibody (23GB6445)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553052
Description
GRB7 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with epidermal growth factor receptor (EGFR) and ephrin receptors. The protein plays a role in the integrin signaling pathway and cell migration by binding with focal adhesion kinase (FAK). Alternative splicing results in multiple transcript variants encoding different isoforms, although the full-length natures of only two of the variants have been determined to date.Chemical Identifiers
| GRB7 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| GRB7 | |
| B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352 | |
| GRB7 | |
| Primary | |
| Affinity chromatography |
| 23GB6445 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14451 | |
| 2886 | |
| A synthesized peptide derived from human GRB7 (1-100AA). | |
| Antibody |
Specifications
| GRB7 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GRB7 | |
| B47; Epidermal growth factor receptor GRB-7; Grb7; GRB7 adapter protein; growth factor receptor binding protein GRB7; growth factor receptor bound protein 7; growth factor receptor-bound protein 7; Growth factor receptor-bound protein 7 (GRB7 adapter protein) (Epidermal growth factor receptor GRB-7); mKIAA4028; OTTHUMP00000164352 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2886 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6445 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14451 | |
| GRB7 | |
| A synthesized peptide derived from human GRB7 (1-100AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |