missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101612 Supplier Invitrogen™ Supplier No. PA5101612
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GRB14.

The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. This protein likely has an inhibitory effect on receptor tyrosine kinase signaling and, in particular, on insulin receptor signaling. This gene may play a role in signaling pathways that regulate growth and metabolism. Transcript variants have been reported for this gene, but their full-length natures have not been determined to date.

Specifications

Antigen GRB14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GRB14
Gene Accession No. O88900, Q14449, Q9JLM9
Gene Alias AI505286; GRB14; GRB14 adapter protein; growth factor receptor bound protein 14; growth factor receptor-bound protein 14
Gene Symbols GRB14
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRB14(Accession Q14449), corresponding to amino acid residues F90-K140.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2888, 50915, 58844
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less