missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GRASP65 Recombinant Rabbit Monoclonal Antibody (23GB5580)
GREENER_CHOICE

Catalog No. MA553902
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553902 100 μL
MA553903 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553902 Supplier Invitrogen™ Supplier No. MA553902
Only null left

Rabbit Recombinant Monoclonal Antibody

GRASP65 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.

Specifications

Antigen GRASP65
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5580
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GORASP1
Gene Accession No. Q9BQQ3
Gene Alias 5430411C10Rik; Golgi peripheral membrane protein p65; Golgi phosphoprotein 5; Golgi reassembly and stacking protein 1; golgi reassembly stacking protein 1; golgi reassembly stacking protein 1, 65kDa; Golgi reassembly-stacking protein 1; Golgi reassembly-stacking protein of 65 kDa; GOLPH5; Gorasp1; GRASP65; P65
Gene Symbols GORASP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GRASP65 (400-440AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less