Learn More
Graphite powder, synthetic, -20+100 mesh, 99% C, 0.2% max Ash, Thermo Scientific Chemicals
Description
Graphite powder is basically used for casting, polishing purposes and molding. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and moderator in nuclear piles. It is used in the preparation of single sheet graphene nanoflakes and graphite based magneto rheological elastomers. It is also employed as a free standing film like cantilever sensor in piezoelectric device and additionally used as a catalyst for wet peroxide oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Mesh Size | −20+100 Mesh |
| Quantity | 250 g |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.9% |
| Physical Form | Powder |
| Assay | (metals basis) |
| Chemical Name or Material | Graphite powder, Synthetic |
Safety and Handling
| Hazard Category | H319-H335 |
| Hazard Statement | GHS H Statement H319-H335 Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
| EINECSNumber | 231-955-3 |
| RTECSNumber | MD9659600 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only