Learn More
Graphite powder, natural, universal grade, -200 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040799TB
Description
Used in aircraft manufacturing industries, defense material, automotive industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Graphite powder | |
| 7782-42-5 | |
| −200 Mesh | |
| Odorless | |
| C | |
| 113 g | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| carbon | |
| 5462310 | |
| 99.9995% | |
| Powder |
| Natural | |
| Black | |
| UCP-1-U grade, ultra 'F' purity | |
| (metals basis) | |
| MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
| UN1325 | |
| Insoluble. | |
| [C] | |
| 12.01 | |
| CHEBI:27594 | |
| Universal |
Chemical Identifiers
| 7782-42-5 | |
| 12.01 | |
| OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
| 5462310 | |
| carbon |
| C | |
| MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
| graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
| CHEBI:27594 | |
| [C] |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 231-955-3
RTECSNumber : MD9659600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only