missing translation for 'onlineSavingsMsg'
Learn More

Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7049572
Change view
Click to view available options
Quantity:
28 g
113 g
227 g
454 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14735TA 28 g
AA14735TB 113 g
AA14735TC 227 g
AA14735TD 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA14735TA Supplier Thermo Scientific Chemicals Supplier No. 014735TA
Only null left

Graphite powder is used to make crucibles, lead pencils, electrodes, lubricants, stove polish, brake linings, batteries, glass manufacture, conductive coatings, and for foundry facings. It is used in the synthesis of single sheet graphene nanoflakes and graphite based magnetorheological elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
Mesh Size −100 Mesh
Odor Odorless
Quantity 28 g
UN Number UN1325
Solubility Information Insoluble in water.
Percent Purity 99.9995%
Grade UCP-1 grade, ultra 'F' purity
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Graphite powder, Natural
Show More Show Less
Hazard Category H228-H319-H335
Hazard Statement GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 231-955-3
RTECSNumber MD9659600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only