missing translation for 'onlineSavingsMsg'
Learn More

Graphite foil, 0.4mm (0.015in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. AA42952VA
Change view
Click to view available options
Quantity:
15 x 15 cm
15 x 60 cm
15 x 250 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42952VA 15 x 15 cm
AA42952CG 15 x 60 cm
AA42952VU 15 x 250 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42952VA Supplier Thermo Scientific Chemicals Supplier No. 042952VA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-42-5
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Assay Percent Range 99.8%, metal basis
Quantity 15 x 15 cm
Physical Form Foil
Chemical Name or Material Graphite
EINECSNumber 231-955-3
RTECSNumber MD9659600
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only