missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granzyme K Monoclonal Antibody (C3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566059
Description
Granzyme K Monoclonal Antibody for Western Blot, IHC (P)
Granzyme K (GrK) belongs to a family of trypsin-like serine proteases localised in the cytotoxic granules of activated T cells and NK cells. It encodes a 28 kDa serine protease whose gene is located on chromosome 5q11-12 close to the granzyme A-encoding gene. Like granzyme A, it has a trypsin-like specifity cleaving at the basic residues arginine and lysine. Granzyme K triggers rapid cell death independently of caspase activation with single-stranded DNA nicks and is primarily expressed in thymus, lung, spleen and peripheral blood leukocytes.Specifications
| Granzyme K | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35205, P49864 | |
| GZMK | |
| Recombinant Granzyme K (GZMK) (Gln44-Val227). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C3 | |
| Unconjugated | |
| GZMK | |
| Fragmentin-3; granzyme 3; granzyme K; granzyme K (granzyme 3; tryptase II); granzyme K (serine protease, granzyme 3; tryptase II); granzyme-3; GrK; GZMK; NK-Tryp-2; NK-tryptase-2; RNK-Tryp-2; TRYP2; tryptase II | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14945, 29165 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |