Learn More
Invitrogen™ Mouse Granzyme B ELISA Kit
ELISA
Supplier: Invitrogen™ BMS6029TEN
Description
The Mouse Granzyme B ELISA quantitates Ms Granzyme B in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Granzyme B. Principle of the method The Mouse Granzyme B solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.Specifications
| P04187 | |
| 1.6 pg/mL | |
| HRP | |
| ELISA | |
| Mouse | |
| AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3EAI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3E; AI553453; CCP-1/C11; CCP1; Ctla-1; Ctla1; GZB | |
| GZMB | |
| 3.5% | |
| Biotin | |
| 10 x 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Granzyme B | |
| 3 hr 30 min |
| 7.8-500 pg/mL | |
| Aluminum pouch(es) with a Microplate coated with monoclonal antibody to mouse Granzyme B Biotin-Conjugate anti-mouse Granzyme B antibody Streptavidin-HRP Mouse Granzyme B Standard lyophilized, 1000 pg/mL upon reconstitution Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Sample Diluent Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Blue-Dye Green-Dye Red-Dye Adhesive Films |
|
| Sandwich ELISA Kit | |
| Plasma, Serum, Supernatant | |
| Absorbance Microplate Reader | |
| 14939 | |
| 4.1% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| CTLA-1, Cytotoxic cell protease 1, CCP1, Fragmentin-2 | |
| RUO | |
| 2-8°C | |
| 1 hr 20 min |