missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Granulins Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527275
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527275 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527275 Supplier Invitrogen™ Supplier No. PA527275
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse kidney, rat kidney. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PCDGF is a glycoprotein and growth factor involved in the regulation of mitosis, early embryonic development, cell division, cell migration, cell survival, and hypothalamic differentiation. PCDGF is also known as progranulin because it is the precursor for smaller proteins known as epithelins or granulins.

Specifications

Antigen Granulins
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene GRN
Gene Accession No. P28799
Gene Alias acrogranin; acrogranulin; CLN11; epithelin; Epithelin precursor; Epithelin/granulin precursor; Epithelin-1; Epithelin-2; GEP; Glycoprotein of 88 Kda; GP88; granulin; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; granulins; Grn; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; PGRN; Proepithelin; progranulin
Gene Symbols GRN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Granulins. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2896
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less