Learn More
Graduated Cylinders
With embossed calibrations for easy measuring.
Supplier:  Customer Channels Group (Ccg) 78200
 View more versions of this product
Catalog No. S95592
Only null left
Explore available promotions
Description
Description
  • Durable, transparent polypropylene
  • Wide, stable bases
  • Chemical and acid resistant
  • Autoclavable (dry heat up to 120°C [248°F], wet heat up to 80°C [176°F])
  • Microwavable
Order Info
Available individually or as seven-piece set.
Specifications
Specifications
Polypropylene
120°C
Graduated Cylinder
10, 25, 50, 100, 250, 500, 1000 mL Cylinders
Wide
Product Suggestions
Product Suggestions