missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149856
Description
GPX6 Polyclonal Antibody for Western Blot, ICC/IF, IHC
The protein encoded by this gene belongs to the glutathione peroxidase family, members of which catalyze the reduction of hydrogen peroxide, organic hydroperoxides and lipid hydroperoxides, and thereby protect cells against oxidative damage. Several isozymes of this gene family exist in vertebrates, which vary in cellular location and substrate specificity. Expression of this gene has been observed in embryos and olfactory epithelium; however, the exact function of this gene is not known. This isozyme is a selenoprotein in humans, containing the rare amino acid selenocysteine (Sec) at its active site. Sec is encoded by the UGA codon, which normally signals translation termination. The 3' UTRs of selenoprotein mRNAs contain a conserved stem-loop structure, designated the Sec insertion sequence (SECIS) element, that is necessary for the recognition of UGA as a Sec codon, rather than as a stop signal. The orthologs of this gene in mouse and rat (and some other species) contain a cysteine (Cys) residue in place of the Sec residue, and their corresponding mRNAs lack SECIS element.Specifications
| GPX6 | |
| Polyclonal | |
| Unconjugated | |
| Gpx6 | |
| Glutathione Peroxidase 6; Glutathione Peroxidase 6 Olfactory | |
| Rabbit | |
| Protein A | |
| RUO | |
| 259233, 75512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q64625, Q91WR8 | |
| Gpx6 | |
| Recombinant Glutathione Peroxidase 6, Olfactory (GPX6) (Gln20-Gln221). | |
| 200 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody |