missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPSN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113868 Supplier Invitrogen™ Supplier No. PA5113868

Rabbit Polyclonal Antibody

Immunogen sequence: LRPAGSKTRK IPYPTKNPFT WLFLLVSCPN YTYEVGSWIG FAIMTQCLPV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.

Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM].

Specifications

Antigen GPSN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TECR
Gene Accession No. Q9NZ01
Gene Alias 2410016D23Rik; A230102P12Rik; AI173355; D17Ertd178e; glycoprotein, synaptic 2; GPSN2; hypothetical protein LOC614105; MRT14; neuroprotective protein 13; SC2; Synaptic glycoprotein SC2; Tecr; TER; trans-2,3-enoyl-CoA reductase; trans-2,3-enoyl-CoA reductase; very-long-chain enoyl-CoA reductase; very-long-chain enoyl-CoA reductase
Gene Symbols TECR
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human GPSN2 (aa 218-267).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9524
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less