missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01675647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01675647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01675647 Supplier Invitrogen™ Supplier No. PA141307
Only null left

Rabbit Polyclonal Antibody

This antibody is predicted to react with xenopus based on sequence homology. Immunogen sequence homology: Gorilla (94%).

GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.

Specifications

Antigen GPR83
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene GPR83
Gene Accession No. P30731, Q9NYM4, Q9TTQ9
Gene Alias AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82
Gene Symbols GPR83
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR83.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104968467, 10888, 140595, 14608, 403699, 428093
Target Species Human, Mouse, Rat, Canine, Monkey, Bovine, Chicken
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less