missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR77 Recombinant Rabbit Monoclonal Antibody (HL4182)
GREENER_CHOICE

Catalog No. MA568427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA568427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA568427 Supplier Invitrogen™ Supplier No. MA568427
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

GPR77 Recombinant Monoclonal Antibody for Western Blot

C5AR2 is an orphan member that belongs to anaphylatoxin receptor family. It is a seven transmembrane G-protein-coupled receptor consisting of an N-terminal N-glycosylation site, several conserved cysteines and several potential C-terminal phosphorylation sites. It binds to C5a and C5a-des-Arg and serves as a highly regulated scavenger receptor with a negative modulatory influence on C5aR-mediated inflammation. Northern Blot analysis suggests it is co- expressed along with C5aR in different tissues and cells including neutrophils/macrophages, mast cells, dendritic cells, brain, lung, heart, kidney, liver, ovary and testis.

Specifications

Antigen GPR77
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL4182
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene C5AR2
Gene Accession No. Q9P296
Gene Alias C5a anaphylatoxin chemotactic receptor 2, C5a anaphylatoxin chemotactic receptor C5L2, C5AR2, C5L2, Complement component 5a receptor 2, G protein-coupled receptor 77, G protein-coupled receptor C5L2, GPF77, GPR77, G-protein coupled receptor 77
Gene Symbols C5AR2
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Intracellular domain of human C5L2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27202
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less