missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR35 Monoclonal Antibody (C3), Invitrogen™
Description
Acts as a receptor for kynurenic acid, an intermediate in the tryptophan metabolic pathway. The activity of this receptor is mediated by G-proteins that elicit calcium mobilization and inositol phosphate production through G(qi/o) proteins. [UniProt].
Specifications
Specifications
| Antigen | GPR35 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | C3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | GPR35 |
| Gene Accession No. | Q9HC97 |
| Gene Alias | G protein-coupled receptor 35; Gpr35; G-protein coupled receptor 35; KPG_007; KYNA receptor; kynurenic acid receptor |
| Show More |