missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR30 Recombinant Rabbit Monoclonal Antibody (20H15L21)

Catalog No. 703480
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
703480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 703480 Supplier Invitrogen™ Supplier No. 703480
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

This antibody is predicted to react with Mouse, Rat.

This gene is a member of the G-protein coupled receptor 1 family and encodes a multi-pass membrane protein that localizes to the endoplasmic reticulum. The protein binds estrogen, resulting in intracellular calcium mobilization and synthesis of phosphatidylinositol 3,4,5-trisphosphate in the nucleus. This protein therefore plays a role in the rapid nongenomic signaling events widely observed following stimulation of cells and tissues with estrogen. Alternate transcriptional splice variants which encode the same protein have been characterized.

Specifications

Antigen GPR30
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 20H15L21
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene GPER1
Gene Accession No. Q99527
Gene Alias 6330420K13Rik; CEPR; Ceprl; chemoattractant receptor-like 2; chemokine receptor-like 2; CMKRL2; constitutively expressed peptide-like receptor; constitutively expressed peptide-like receptor like; DRY12; FEG-1; Flow-induced endothelial G-protein coupled receptor 1; G protein-coupled estrogen receptor 1; G protein-coupled receptor 30; GPCR-Br; GPER; GPER1; Gpr30; GPR41; G-protein coupled estrogen receptor 1; G-protein coupled receptor 30; G-protein coupled receptor 41; heptahelix receptor; IL8-related receptor DRY12; LERGU; LERGU2; leucine rich protein in GPR30 3'UTR; LyGPR; Lymphocyte-derived G-protein coupled receptor; membrane estrogen receptor; mER
Gene Symbols GPER1
Host Species Rabbit
Immunogen Peptides corresponding to human GPR30 [1) aa353-aa375 2) aa2-aa19].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less