missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR177 Monoclonal Antibody (A7C2)

Description
GPR177 Monoclonal Antibody for Western Blot, ICC/IF
Regulates Wnt proteins sorting and secretion in a feedback regulatory mechanism. This reciprocal interaction plays a key role in the regulation of expression, subcellular location, binding and organelle-specific association of Wnt proteins. Plays also an important role in establishment of the anterior-posterior body axis formation during development. [UniProt].
Specifications
Specifications
| Antigen | GPR177 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A7C2 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | WLS |
| Gene Accession No. | Q5T9L3, Q6DID7, Q6P689 |
| Gene Alias | 5031439A09Rik; AI173978; AI987742; C1orf139; evenness interrupted homolog; EVI; G protein-coupled receptor 177; Gpr177; Integral membrane protein GPR177; mig-14; MRP; Protein evenness interrupted homolog; protein wntless homolog; Putative NF-kappa-B-activating protein 373; putative NFkB activating protein 373; UNQ85/PRO18667; WLS; Wnt ligand secretion mediator; wntless homolog; wntless homolog (Drosophila); wntless Wnt ligand secretion mediator |
| Show More |