missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR174 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104386
Description
Antibody detects endogenous levels of total GPR174.
G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR174 (G protein-coupled receptor 174), also known as FKSG79, is a 333 amino acid multi-pass membrane protein that belongs to the G-protein coupled receptor 1 family and is thought to function as a receptor for G protein-coupled purines.Specifications
| GPR174 | |
| Polyclonal | |
| Unconjugated | |
| GPR174 | |
| FKSG79; G protein-coupled receptor 174; Gm376; GPCR17; GPR174; Jeg18; LYPSR3; probable G-protein coupled receptor 174; putative purinergic receptor FKSG79; RGD1564689 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84636 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BXC1 | |
| GPR174 | |
| A synthesized peptide derived from human GPR174(Accession Q9BXC1), corresponding to amino acid residues K132-S182. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |